LIFH93CB08D9 Life Chemicals Inc.

(Z)-5-bromo-N'-hydroxypyrimidine-2-carboximidamide

  • MDL number MFCD30483074
  • Cas Number : 1854110-32-9
  • Molecular Weight : 217.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (5-bromopyrimidin-2-yl)(nitroso)methanamine
Inchi Code 1S/C5H5BrN4O/c6-3-1-8-5(9-2-3)4(7)10-11/h1-2,4H,7H2
Inchi Key VHZLMDVABIZODB-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart