LIFH93CB08DB Life Chemicals Inc.

2-(3,3-difluoroazetidin-1-yl)acetic acid hydrochloride

  • MDL number MFCD31625834
  • Cas Number : 2320574-31-8
  • Molecular Weight : 187.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(3,3-difluoroazetidin-1-yl)acetic acid hydrochloride
Inchi Code 1S/C5H7F2NO2.ClH/c6-5(7)2-8(3-5)1-4(9)10;/h1-3H2,(H,9,10);1H
Inchi Key KLTXLTIAWIEKOP-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart