LIFH93CB08DE Life Chemicals Inc.

trans-4-(difluoromethyl)cyclohexane-1-carboxylic acid

  • MDL number MFCD29046547
  • Cas Number : 1115331-05-9
  • Molecular Weight : 178.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1r,4r)-4-(difluoromethyl)cyclohexane-1-carboxylic acid
Inchi Code 1S/C8H12F2O2/c9-7(10)5-1-3-6(4-2-5)8(11)12/h5-7H,1-4H2,(H,11,12)/t5-,6-
Inchi Key YYQCHCJHDZCDSW-IZLXSQMJSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart