LIFH93CB08E4 Life Chemicals Inc.

1-cyclobutylpiperidin-3-amine

  • MDL number MFCD19611359
  • Cas Number : 1342397-12-9
  • Molecular Weight : 154.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-cyclobutyl-3-piperidinamine
Inchi Code 1S/C9H18N2/c10-8-3-2-6-11(7-8)9-4-1-5-9/h8-9H,1-7,10H2
Inchi Key OGSPMLPOTYMZBP-UHFFFAOYSA-N

Description

Purity 95%
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart