LIFH93CB08E5 Life Chemicals Inc.

1-ethyl-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD08704772
  • Cas Number : 4778-76-1
  • Molecular Weight : 139.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-ethyl-1H-pyrrole-2-carboxylic acid
Inchi Code 1S/C7H9NO2/c1-2-8-5-3-4-6(8)7(9)10/h3-5H,2H2,1H3,(H,9,10)
Inchi Key XCOJUFQICAEQEP-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart