LIFH93CB08E9 Life Chemicals Inc.

1-(2-bromoethyl)-5-fluoro-1H-indole

  • MDL number MFCD31672243
  • Cas Number : 1865988-59-5
  • Molecular Weight : 242.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2-bromoethyl)-5-fluoro-1H-indole
Inchi Code 1S/C10H9BrFN/c11-4-6-13-5-3-8-7-9(12)1-2-10(8)13/h1-3,5,7H,4,6H2
Inchi Key QISUTVLUPHJMDU-UHFFFAOYSA-N

Description

Purity 95%
Physical Form oil
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart