LIFH93CB08F4 Life Chemicals Inc.

1,3-bis(difluoromethyl)benzene

  • MDL number MFCD29767780
  • Cas Number : 936330-10-8
  • Molecular Weight : 178.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1,3-bis(difluoromethyl)benzene
Inchi Code 1S/C8H6F4/c9-7(10)5-2-1-3-6(4-5)8(11)12/h1-4,7-8H
Inchi Key ICPZMZNMPALADE-UHFFFAOYSA-N

Description

Purity 95%
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart