LIFH93CB08F8 Life Chemicals Inc.

3-(2-aminoethyl)-3H,4H-thieno[3,2-d]pyrimidin-4-one dihydrochloride

  • MDL number MFCD31736210
  • Cas Number : 2247102-69-6
  • Molecular Weight : 268.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(2-aminoethyl)thieno[3,2-d]pyrimidin-4(3H)-one dihydrochloride
Inchi Code 1S/C8H9N3OS.2ClH/c9-2-3-11-5-10-6-1-4-13-7(6)8(11)12;;/h1,4-5H,2-3,9H2;2*1H
Inchi Key IBFGWEQERDCICC-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart