LIFH93CB08FE Life Chemicals Inc.

3-(2-aminoethyl)aniline hydrochloride

  • MDL number MFCD07371488
  • Cas Number : 1217099-81-4
  • Molecular Weight : 172.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(2-aminoethyl)aniline hydrochloride
Inchi Code 1S/C8H12N2.ClH/c9-5-4-7-2-1-3-8(10)6-7;/h1-3,6H,4-5,9-10H2;1H
Inchi Key HBBWQVPXGBHVPE-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart