LIFH93CB0901 Life Chemicals Inc.

3-bromo-5-iodo-2,4,6-trimethylpyridine

  • Cas Number : 29976-61-2
  • Molecular Weight : 325.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-5-iodo-2,4,6-trimethylpyridine

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart