LIFH93CB0903 Life Chemicals Inc.

2-bromo-5-iodo-4,6-dimethylpyridin-3-amine

  • Cas Number : 2415263-12-4
  • Molecular Weight : 326.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-5-iodo-4,6-dimethylpyridin-3-amine

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart