LIFH93CB0908 Life Chemicals Inc.

3-(propan-2-yl)-5H,6H,7H,8H-imidazo[1,5-a]pyridine-1-carboxylic acid

  • MDL number MFCD26134830
  • Cas Number : 1510467-54-5
  • Molecular Weight : 208.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-isopropyl-5,6,7,8-tetrahydroimidazo[1,5-a]pyridine-1-carboxylic acid
Inchi Code 1S/C11H16N2O2/c1-7(2)10-12-9(11(14)15)8-5-3-4-6-13(8)10/h7H,3-6H2,1-2H3,(H,14,15)
Inchi Key GZMGYMIDULBJOF-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart