LIFH93CB0909 Life Chemicals Inc.

2-(5-amino-3-cyclopropyl-1H-pyrazol-1-yl)-6-propyl-3,4-dihydropyrimidin-4-one

  • MDL number MFCD16631890
  • Cas Number : 1207026-00-3
  • Molecular Weight : 259.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(5-amino-3-cyclopropyl-1H-pyrazol-1-yl)-6-propylpyrimidin-4-ol
Inchi Code 1S/C13H17N5O/c1-2-3-9-6-12(19)16-13(15-9)18-11(14)7-10(17-18)8-4-5-8/h6-8H,2-5,14H2,1H3,(H,15,16,19)
Inchi Key CBYPRDWFXKAONH-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart