LIFH93CB090E Life Chemicals Inc.

2,4-difluoro-1-(pentafluoro-lambda6-sulfanyl)benzene

  • Cas Number : 1522408-96-3
  • Molecular Weight : 240.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,4-difluorophenyl)pentafluoro-lambda6-sulfane

Description

Purity 95%
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart