LIFH93CB0910 Life Chemicals Inc.

1-[(3-chloro-5-cyanophenyl)sulfanyl]-N,N-dimethylformamide

  • Cas Number : 1978338-13-4
  • Molecular Weight : 240.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name S-(3-chloro-5-cyanophenyl) dimethylcarbamothioate
Inchi Code 1S/C10H9ClN2OS/c1-13(2)10(14)15-9-4-7(6-12)3-8(11)5-9/h3-5H,1-2H3
Inchi Key SDCVXLRPELRZIA-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart