LIFH93CB0976 Life Chemicals Inc.

methyl 3-(5-methylthiophen-2-yl)-1H-pyrazole-5-carboxylate

  • MDL number MFCD09143219
  • Cas Number : 1025405-27-9
  • Molecular Weight : 222.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart