LIFH93CB0982 Life Chemicals Inc.

1-(3,3-difluorocyclobutanecarbonyl)piperidin-3-ol

  • Cas Number : 1857258-56-0
  • Molecular Weight : 219.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,3-difluorocyclobutyl)(3-hydroxypiperidin-1-yl)methanone
Inchi Code 1S/C10H15F2NO2/c11-10(12)4-7(5-10)9(15)13-3-1-2-8(14)6-13/h7-8,14H,1-6H2
Inchi Key YVGIIJXTXMCUTJ-UHFFFAOYSA-N

Description

Purity 95%
Physical Form oil
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart