LIFH93DAE03F Life Chemicals Inc.

(1-cyclopentylpiperidin-4-yl)methanol

  • MDL number MFCD16162898
  • Cas Number : 953026-60-3
  • Molecular Weight : 183.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1-cyclopentyl-4-piperidinyl)methanol
Inchi Code 1S/C11H21NO/c13-9-10-5-7-12(8-6-10)11-3-1-2-4-11/h10-11,13H,1-9H2
Inchi Key RNZVVXVTFVPCBK-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart