LIFH93DAE040 Life Chemicals Inc.

methyl 1-methyl-3-(pyridin-2-yl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD30008869
  • Cas Number : 2098141-76-3
  • Molecular Weight : 217.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 1-methyl-3-(pyridin-2-yl)-1H-pyrazole-4-carboxylate
Inchi Code 1S/C11H11N3O2/c1-14-7-8(11(15)16-2)10(13-14)9-5-3-4-6-12-9/h3-7H,1-2H3
Inchi Key XBRBWSXTDUBEDA-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart