LIFH93DAE04F Life Chemicals Inc.

6-fluoro-1-methyl-2-oxo-1,2-dihydroquinoline-3-carboxylic acid

  • MDL number MFCD26625682
  • Cas Number : 1498999-88-4
  • Molecular Weight : 221.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-fluoro-1-methyl-2-oxo-1,2-dihydroquinoline-3-carboxylic acid
Inchi Code 1S/C11H8FNO3/c1-13-9-3-2-7(12)4-6(9)5-8(10(13)14)11(15)16/h2-5H,1H3,(H,15,16)
Inchi Key NAJIAFSVLHUQLX-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart