LIFH944550ED Life Chemicals Inc.

2-chloro-4-fluoro-5-iodobenzoic acid

  • MDL number MFCD24107204
  • Cas Number : 264927-52-8
  • Molecular Weight : 300.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart