LIVH3031A435 Liverpool ChiroChem

4-(2,3-dichlorophenyl)piperidine hydrochloride

  • Cas Number : 1354030-49-1
  • Molecular Weight : 230.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Salt Data HCl
Country of Origin United Kingdom
Product Link www.liverpoolchirochem.com/A0225

Safety Information

hcodes GHS07
MSDS www.liverpoolchirochem.com/A0225
Availability details for
Quantity
  • Availability
  • Add to Cart