LIVH3031A585 Liverpool ChiroChem

2-(Tetrahydrofuran-3-yl)acetic acid

  • Cas Number : 138498-97-2
  • Molecular Weight : 130.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link www.liverpoolchirochem.com/BC0222

Safety Information

hcodes GHS07
MSDS www.liverpoolchirochem.com/BC0222
Availability details for
Quantity
  • Availability
  • Add to Cart