LIVH30324EA2 Liverpool ChiroChem

tert-butyl (R)-2-methylpiperazine-1-carboxylate

  • Cas Number : 170033-47-3
  • Molecular Weight : 200.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link www.liverpoolchirochem.com/A1480

Safety Information

hcodes GHS07
MSDS www.liverpoolchirochem.com/A1480
Availability details for
Quantity
  • Availability
  • Add to Cart