LIVH93E9FB3B Liverpool ChiroChem

(2S,4R)-1-(tert-butoxycarbonyl)-4-(2-fluorophenyl)pyrrolidine-2-carboxylic acid

  • Molecular Weight : 309.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/A8942?search=A8942

Safety Information

MSDS https://www.liverpoolchirochem.com/A8942?search=A8942
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart