LIVH93E9FB44 Liverpool ChiroChem

(2S,4R)-1-(((9H-fluoren-9-yl)methoxy)carbonyl)-4-(4-fluorophenyl)pyrrolidine-2-carboxylic acid

  • Molecular Weight : 431.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/A9249?search=A9249

Safety Information

MSDS https://www.liverpoolchirochem.com/A9249?search=A9249
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart