LIVH93E9FB5A Liverpool ChiroChem

5-formyl-1H-indazole-3-carbonitrile

  • Cas Number : 1823867-50-0
  • Molecular Weight : 171.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/A0366?search=A0366

Safety Information

MSDS https://www.liverpoolchirochem.com/A0366?search=A0366
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart