LIVH93E9FB63 Liverpool ChiroChem

3-(2-fluorophenyl)-2,2-dimethylpyrrolidine

  • Cas Number : 1250794-27-4
  • Molecular Weight : 193.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/A1468?search=A1468

Safety Information

MSDS https://www.liverpoolchirochem.com/A1468?search=A1468
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart