LIVH93E9FB6E Liverpool ChiroChem

2-amino-1-(2,2-difluorobenzo[d][1,3]dioxol-5-yl)ethan-1-one

  • Cas Number : 1380045-08-8
  • Molecular Weight : 215.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/A2144?search=A2144

Safety Information

MSDS https://www.liverpoolchirochem.com/A2144?search=A2144
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart