LIVH93E9FB8A Liverpool ChiroChem

6-bromo-1-methyl-1H-indazole-3-carbaldehyde

  • Cas Number : 1781630-54-3
  • Molecular Weight : 239.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/A3483?search=A3483

Safety Information

MSDS https://www.liverpoolchirochem.com/A3483?search=A3483
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart