LIVH93E9FB8C Liverpool ChiroChem

1-(tert-butyl) 2-methyl (R)-4-oxopiperidine-1,2-dicarboxylate

  • Cas Number : 1799811-83-8
  • Molecular Weight : 257.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/A3554?search=A3554

Safety Information

MSDS https://www.liverpoolchirochem.com/A3554?search=A3554
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart