LIVH93E9FB94 Liverpool ChiroChem

5,5-dimethylpiperidine-2,4-dione

  • Cas Number : 118263-81-3
  • Molecular Weight : 141.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/A4007?search=A4007

Safety Information

MSDS https://www.liverpoolchirochem.com/A4007?search=A4007
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart