LIVH93E9FBB1 Liverpool ChiroChem

1-(1-Benzyl-5-methoxy-1H-indol-3-yl)-2-chloroethan-1-one

  • Cas Number : 1824267-13-1
  • Molecular Weight : 313.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/A4998?search=A4998

Safety Information

MSDS https://www.liverpoolchirochem.com/A4998?search=A4998
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart