LIVH93E9FBB5 Liverpool ChiroChem

(S)-1-(4-chlorophenyl)ethan-1-amine

  • Cas Number : 4187-56-8
  • Molecular Weight : 155.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/A5137?search=A5137

Safety Information

MSDS https://www.liverpoolchirochem.com/A5137?search=A5137
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart