LIVH93E9FBB9 Liverpool ChiroChem

(S)-1-(4-(trifluoromethyl)phenyl)ethan-1-amine

  • Cas Number : 84499-73-0
  • Molecular Weight : 189.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/A5141?search=A5141

Safety Information

MSDS https://www.liverpoolchirochem.com/A5141?search=A5141
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart