LIVH93E9FBCA Liverpool ChiroChem

(8S,9R)-5-fluoro-8-(4-fluorophenyl)-9-(1-methyl-1H-1,2,4-triazol-5-yl)-8,9-dihydro-2H-pyrido[4,3,2-de]phthalazin-3(7H)-one tosylate

  • Cas Number : 1373431-65-2
  • Molecular Weight : 552.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/A5245?search=A5245

Safety Information

MSDS https://www.liverpoolchirochem.com/A5245?search=A5245
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart