LIVH93E9FBD4 Liverpool ChiroChem

4-methoxybenzyl 2-bromoacetate

  • Cas Number : 63353-51-5
  • Molecular Weight : 259.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/A5570?search=A5570

Safety Information

MSDS https://www.liverpoolchirochem.com/A5570?search=A5570
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart