LIVH93E9FBF4 Liverpool ChiroChem

tert-butyl (piperidin-3-ylmethyl)carbamate

  • Cas Number : 142643-29-6
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/A7972?search=A7972

Safety Information

MSDS https://www.liverpoolchirochem.com/A7972?search=A7972
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart