LIVH93E9FBF5 Liverpool ChiroChem

methyl 4-ethyl-2-methyl-1H-imidazole-5-carboxylate

  • Cas Number : 1245644-44-3
  • Molecular Weight : 168.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/A7978?search=A7978

Safety Information

MSDS https://www.liverpoolchirochem.com/A7978?search=A7978
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart