LIVH93E9FBFD Liverpool ChiroChem

1-benzyl-5-((benzyloxy)carbonyl)-2-methylpyridin-1-ium chloride

  • Molecular Weight : 353.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/A8125?search=A8125

Safety Information

MSDS https://www.liverpoolchirochem.com/A8125?search=A8125
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart