LIVH93E9FBFF Liverpool ChiroChem

(1-benzyl-1H-imidazol-5-yl)methanol

  • Cas Number : 80304-50-3
  • Molecular Weight : 188.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/A8190?search=A8190

Safety Information

MSDS https://www.liverpoolchirochem.com/A8190?search=A8190
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart