LIVH93E9FC02 Liverpool ChiroChem

2-amino-N-(quinolin-8-yl)benzenesulfonamide

  • Cas Number : 16082-64-7
  • Molecular Weight : 299.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/A8249?search=A8249

Safety Information

MSDS https://www.liverpoolchirochem.com/A8249?search=A8249
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart