LIVH93E9FC16 Liverpool ChiroChem

(S)-2,3-dihydro-1H-inden-1-amine hydrochloride

  • Cas Number : 32457-23-1
  • Molecular Weight : 169.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl salt
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/A8576?search=A8576

Safety Information

MSDS https://www.liverpoolchirochem.com/A8576?search=A8576
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart