LIVH93E9FC17 Liverpool ChiroChem

(2S,3S)-1-(tert-butoxycarbonyl)-2-methylpyrrolidine-3-carboxylic acid

  • Cas Number : 664364-34-5
  • Molecular Weight : 229.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/A8612?search=A8612

Safety Information

MSDS https://www.liverpoolchirochem.com/A8612?search=A8612
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart