LIVH93E9FC30 Liverpool ChiroChem

5-methyl-1,3,4-thiadiazole-2-carbonitrile

  • Cas Number : 518015-09-3
  • Molecular Weight : 125.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/A0711?search=A0711

Safety Information

MSDS https://www.liverpoolchirochem.com/A0711?search=A0711
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart