LIVH93E9FC3F Liverpool ChiroChem

2-(3-(piperidin-4-yl)phenyl)ethan-1-amine dihydrochloride

  • Molecular Weight : 277.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl salt
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/A10146?search=A10146

Safety Information

MSDS https://www.liverpoolchirochem.com/A10146?search=A10146
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart