LIVH93E9FC41 Liverpool ChiroChem

(R)-3-(1-(tert-butoxycarbonyl)piperidin-2-yl)benzoic acid

  • Molecular Weight : 305.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/A9675?search=A9675

Safety Information

MSDS https://www.liverpoolchirochem.com/A9675?search=A9675
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart