LIVH93E9FC54 Liverpool ChiroChem

cis-4-(((9H-fluoren-9-yl)methoxy)carbonyl)-6-methylmorpholine-2-carboxylic acid

  • Molecular Weight : 367.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/A9725?search=A9725

Safety Information

MSDS https://www.liverpoolchirochem.com/A9725?search=A9725
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart