LIVH93E9FC56 Liverpool ChiroChem

1,8-naphthyridin-2-amine

  • Cas Number : 15992-83-3
  • Molecular Weight : 145.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/A1024?search=A1024

Safety Information

MSDS https://www.liverpoolchirochem.com/A1024?search=A1024
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart