LIVH93E9FC58 Liverpool ChiroChem

(R)-2-(3-bromophenyl)-1,4-oxazepane

  • Cas Number : 1246456-41-6
  • Molecular Weight : 256.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/A10422?search=A10422

Safety Information

MSDS https://www.liverpoolchirochem.com/A10422?search=A10422
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart